Masterflex® Touch-Screen Syringe Pumps
Control flow rate via the user-friendly touch-screen display
  • Flow rates from 5 pL/min to 220 mL/min
  • Full touch-screen enables you to quickly create configurations and recall them later
  • Pump can be placed on its side to provide a smaller footprint with the touch-screen display automatically rotating
  • Accuracy of ±0.3%
These Masterflex® touch-screen syringe pumps offer user-friendly programming and control. All pumps feature an accuracy of ±0.355% and a reproducibility of ±0.05% along with output options RS-232, RS-485, USB, and 15 pin D-sub connector.
$3,466.50 - $5,485.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate Per Syringe (mL/min) Min Flow Rate Per Syringe (picoL/min) Description
Availability Pricing
COMPARE
215.8 3.06 Syringe Pump, Only, Touchscreen Control 74905-50
$3,466.50USD / Each
COMPARE
215.8 3.06 Syringe Pump, Infusion and Withdrawal, Touchscreen Control 74905-52
$4,500.50USD / Each
COMPARE
215.8 3.06 Syringe Pump, Infusion and Withdrawal Programmable, Touchscreen Control
$4,616.00USD / Each
COMPARE
215.8 3.06 Syringe Pump, Push - Pull, Touchscreen Control
$5,069.00USD / Each
COMPARE
215.8 3.06 Syringe Pump, Push - Pull Programable, Touchscreen Control 74905-58
$5,485.00USD / Each