Masterflex® Transfer Tubing, C-Flex®, Opaque White
Meets the demands of various industries
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: General-purpose lab uses requiring high performance and long life.
Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Opaque white.
Certification: Resin meets USP Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -50 to 275°F (-45 to 135°C)
Sterilize: With autoclave, ethylene oxide, or gamma irradiation.
$32.50 - $179.00USD / Pkg of 1
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
13 3/16 5/16
$60.00USD / Pkg of 1
COMPARE
10 3/8 1/2
$83.00USD / Pkg of 1
COMPARE
10 1/2 11/16
$161.00USD / Pkg of 1
COMPARE
30 1/50 1/12
$41.75USD / Pkg of 1
COMPARE
31 1/32 3/32
$32.50USD / Pkg of 1
COMPARE
15 1/16 1/8
$41.00USD / Pkg of 1
COMPARE
14 3/32 5/32
$46.75USD / Pkg of 1
COMPARE
10 1/8 3/16
$48.00USD / Pkg of 1
COMPARE
10 5/32 7/32
$47.00USD / Pkg of 1
COMPARE
9 3/16 1/4
$45.50USD / Pkg of 1
COMPARE
21 1/8 1/4
$44.75USD / Pkg of 1
COMPARE
22 3/16 7/16
$131.00USD / Pkg of 1
COMPARE
12 1/4 3/8
$74.00USD / Pkg of 1
COMPARE
17 1/4 7/16
$93.00USD / Pkg of 1
COMPARE
17 1/4 1/2
$125.00USD / Pkg of 1
COMPARE
10 5/16 7/16
$76.00USD / Pkg of 1
COMPARE
16 5/16 1/2
$97.00USD / Pkg of 1
COMPARE
14 3/8 5/8
$128.00USD / Pkg of 1
COMPARE
10 1/2 3/4
$179.00USD / Pkg of 1
Showing 1- 19 of 19