Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC
Eliminate odor transferring to sample
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Laboratory, food & beverage, pharmaceutical, environmental, and general lab applications.
Characteristics: Translucent, odorless, low water absorption, low extractables
Certified compliant FDA compliant, USP Class VI, 3A compliant, Reach compliant and RoHS compliant.
Temperature range: -58 to 165°F (-50 to 74°C)
Sterilize: Autoclave or ethylene oxide.
$37.75 - $241.00USD / of 1
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/8 1/4 1/16
$37.75USD / of 1
COMPARE
3/16 5/16 1/16
$51.00USD / of 1
COMPARE
1/4 3/8 1/16
$62.00USD / of 1
COMPARE
1/4 1/2 1/8
$76.50USD / of 1
COMPARE
5/16 7/16 1/16
$68.50USD / of 1
COMPARE
3/8 1/2 1/16
$82.00USD / of 1
COMPARE
3/8 3/8 1/8
$157.00USD / of 1
COMPARE
1/2 5/8 1/16
$102.00USD / of 1
COMPARE
1/2 3/4 1/8
$103.00USD / of 1
COMPARE
5/8 7/8 1/8
$158.00USD / of 1
COMPARE
3/4 1 1/8
$165.00USD / of 1
COMPARE
1 1 1/4 1/8
$241.00USD / of 1
Showing 1- 12 of 12