Masterflex® Transfer Tubing, Ester-Based Polyurethane
Worry-free petroleum-based transfer
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
The tubes are ideal for high-purity, general laboratory, petroleum-based transfer. Characteristics include plasticizer-free, low extractables, good at low temperatures, good resistance to petroleum-based chemicals, and high tensile strength. Not recommended for use with aqueous solutions. Temperature range is -70 to 185°F (-56 to 85°C). Can be sterilized with ethylene oxide.
$47.75 - $892.00USD / Pkg of 1
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
100 1/16 1/8
$47.75USD / Pkg of 1
COMPARE
72 1/8 3/16
$59.50USD / Pkg of 1
COMPARE
100 1/8 1/4
$110.00USD / Pkg of 1
COMPARE
27 3/16 1/4
$74.50USD / Pkg of 1
COMPARE
74 3/16 5/16
$134.00USD / Pkg of 1
COMPARE
33 1/4 5/16
$138.00USD / Pkg of 1
COMPARE
67 1/4 3/8
$161.00USD / Pkg of 1
COMPARE
74 1/4 7/16
$291.00USD / Pkg of 1
COMPARE
100 1/4 1/2
$389.00USD / Pkg of 1
COMPARE
65 5/16 7/16
$236.00USD / Pkg of 1
COMPARE
59 3/8 1/2
$202.00USD / Pkg of 1
COMPARE
84 3/8 9/16
$276.00USD / Pkg of 1
COMPARE
100 3/8 5/8
$387.00USD / Pkg of 1
COMPARE
74 7/16 5/8
$359.00USD / Pkg of 1
COMPARE
36 1/2 5/8
$259.00USD / Pkg of 1
COMPARE
60 1/2 3/4
$502.00USD / Pkg of 1
COMPARE
54 5/8 13/16
$482.00USD / Pkg of 1
COMPARE
60 5/8 7/8
$552.00USD / Pkg of 1
COMPARE
51 3/4 1
$606.00USD / Pkg of 1
COMPARE
40 1 1 1/4
$892.00USD / Pkg of 1
Showing 1- 20 of 20