Masterflex® Transfer Tubing, High-Purity PFA-450
Won't crack under stress
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Pharmaceutical, biotechnology, environmental, semiconductor processing, food and beverage, autoanalyzer, and nuclear applications.
Characteristics: Superior surface smoothness and clarity, low extractables with deionized water and acids, high resistance to stress and cracking. Translucent.
Resin certification: Resin is FDA compliant.
Tubing is manufactured according to GMP.
Temperature range: -320 to 500°F (-195 to 260°C)
Sterilize: With autoclave, ethylene oxide, gamma irradiation, and using dry heat.
$68.50 - $890.00USD / Pkg of 1
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
592 1/16 1/8
$68.50USD / Pkg of 1
COMPARE
387 1/8 3/16
$131.00USD / Pkg of 1
COMPARE
600 1/8 1/4
$217.00USD / Pkg of 1
COMPARE
421 5/32 1/4
$190.00USD / Pkg of 1
COMPARE
278 3/16 1/4
$134.00USD / Pkg of 1
COMPARE
388 1/4 3/8
$287.00USD / Pkg of 1
COMPARE
283 3/8 1/2
$418.00USD / Pkg of 1
COMPARE
220 1/2 5/8
$625.00USD / Pkg of 1
COMPARE
150 5/8 3/4
$694.00USD / Pkg of 1
COMPARE
140 3/4 7/8
$810.00USD / Pkg of 1
COMPARE
133 7/8 1
$890.00USD / Pkg of 1
Showing 1- 11 of 11