Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF
Excellent chemical resistance and UV stable
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: High purity and temperature, UV sensitive, or aggressive chemical fluid transfer.
Characteristics: Excellent chemical resistance, low TOC, rigid tubing works well with compression and over-molded fittings and adapters. Translucent.
Certification: FDA complaint (21 CFR 177.2600), meets USP Class VI requirements, and is made in accordance to cGMP.
Temperature range: -40 to 275°F (-40 to 135°C)
Sterilize: Autoclave, gamma irradiation, or ethylene oxide.
$70.50 - $570.00USD / Pkg
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
451 1/16 1/8
$70.50USD / Pkg
COMPARE
168 1/8 3/16
$99.50USD / Pkg
COMPARE
126 3/16 1/4
$103.00USD / Pkg
COMPARE
101 1/4 5/16
$133.00USD / Pkg
COMPARE
84 5/16 3/8
$144.00USD / Pkg
COMPARE
260 1/8 1/4
$151.00USD / Pkg
COMPARE
174 1/4 3/8
$222.00USD / Pkg
COMPARE
130 3/8 1/2
$289.00USD / Pkg
COMPARE
104 3/8 5/8
$369.00USD / Pkg
COMPARE
87 5/8 3/4
$426.00USD / Pkg
COMPARE
65 7/8 1
$570.00USD / Pkg
Showing 1- 11 of 11