Masterflex® Transfer Tubing, Norprene®
Resists chemicals and UV-light
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
This tubing is ideal for chemical, heat, ozone, and ultraviolet (UV) light resistance applications. Characteristics include nonconductive, heat-sealable, nonaging, nonoxidizing, with superior acid and alkali resistance. They are manufactured according to GMP. Temperature range is -75 to 275°F (-60 to 135°C). Can be sterilized with autoclave.
$55.00 - $444.00USD / Pkg of 1
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
34 1/16 3/16
$56.00USD / Pkg of 1
COMPARE
19 1/8 1/4
$55.00USD / Pkg of 1
COMPARE
34 1/8 3/8
$107.00USD / Pkg of 1
COMPARE
13 3/16 5/16
$61.50USD / Pkg of 1
COMPARE
10 1/4 3/8
$71.50USD / Pkg of 1
COMPARE
15 1/4 7/16
$103.00USD / Pkg of 1
COMPARE
19 1/4 1/2
$159.00USD / Pkg of 1
COMPARE
8 5/16 7/16
$89.00USD / Pkg of 1
COMPARE
12 5/16 1/2
$131.00USD / Pkg of 1
COMPARE
7 3/8 1/2
$96.50USD / Pkg of 1
COMPARE
10 3/8 9/16
$155.00USD / Pkg of 1
COMPARE
13 3/8 5/8
$196.00USD / Pkg of 1
COMPARE
6 7/16 9/16
$172.00USD / Pkg of 1
COMPARE
6 1/2 5/8
$137.00USD / Pkg of 1
COMPARE
8 1/2 11/16
$207.00USD / Pkg of 1
COMPARE
7 5/8 13/16
$285.00USD / Pkg of 1
COMPARE
8 5/8 7/8
$308.00USD / Pkg of 1
COMPARE
7 3/4 1
$347.00USD / Pkg of 1
COMPARE
6 1 1 1/4
$444.00USD / Pkg of 1
Showing 1- 19 of 19