Masterflex® Transfer Tubing, PharMed® BPT
Protect your light-sensitive samples
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165°F (-46 to 73°C). Can be sterilized with ethylene oxide or autoclave.
$93.00 - $524.00USD / Pkg of 1
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
50 1/50 9/64
$93.00USD / Pkg of 1
COMPARE
40 1/32 5/32
$116.00USD / Pkg of 1
COMPARE
40 1/16 1/8
$132.00USD / Pkg of 1
COMPARE
34 1/16 3/16
$143.00USD / Pkg of 1
COMPARE
40 1/8 3/16
$164.00USD / Pkg of 1
COMPARE
19 1/8 1/4
$139.00USD / Pkg of 1
COMPARE
13 3/16 5/16
$144.00USD / Pkg of 1
COMPARE
10 1/4 3/8
$168.00USD / Pkg of 1
COMPARE
19 1/4 1/2
$316.00USD / Pkg of 1
COMPARE
8 5/16 7/16
$187.00USD / Pkg of 1
COMPARE
8 3/8 1/2
$208.00USD / Pkg of 1
COMPARE
13 3/8 5/8
$283.00USD / Pkg of 1
COMPARE
11 1/2 3/4
$342.00USD / Pkg of 1
COMPARE
8 5/8 7/8
$419.00USD / Pkg of 1
COMPARE
7 3/4 1
$524.00USD / Pkg of 1
COMPARE
13 3/32 7/32
$176.00USD / Pkg of 1
Showing 1- 16 of 16