Masterflex® Transfer Tubing, Terminator®
Superior abrasion resistance at high pressure
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Outstanding resistance to heat and ozone
Masterflex® Terminator® transfer tubing is ideal for use with severe service applications due to its superior abrasion resistance. The Buna-N nitrile tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube and tube cover meet RMA Class A high oil resistance and nonconductive at 1000 VDC certification.
$316.00 - $568.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
100 1/4 9/16
$316.00USD / Each
COMPARE
100 1/2 57/64
$568.00USD / Each
COMPARE
50 3/4 1 1/5
$491.00USD / Each
COMPARE
50 1 1 33/64
$513.00USD / Each
COMPARE
25 1 1/2 2 5/64
$525.00USD / Each
COMPARE
25 2 2 41/64
$567.00USD / Each