Masterflex® Transfer Tubing, Tygon® F-4040-A, Fuel and Lubricant
No swelling or embrittlement
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
The Tygon tubing is ideal for fuels and industrial lubricants—gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. Resists most hydrocarbons, embrittlement and swelling, ozone- and UV-resistant, with low-extractability. Tubing is manufactured according to GMP and has a temperature range of -35 to 165°F (-37 to 74°C). Sterilization is not recommended.
$58.00 - $469.00USD / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
52 3/32 3/16
$58.00USD / Pkg of 1
COMPARE
52 1/8 1/4
$72.50USD / Pkg of 1
COMPARE
37 3/16 5/16
$105.00USD / Pkg of 1
COMPARE
30 1/4 3/8
$115.00USD / Pkg of 1
COMPARE
25 5/16 7/16
$146.00USD / Pkg of 1
COMPARE
21 3/8 1/2
$173.00USD / Pkg of 1
COMPARE
19 7/16 9/16
$209.00USD / Pkg of 1
COMPARE
30 1/2 3/4
$370.00USD / Pkg of 1
COMPARE
25 5/8 7/8
$437.00USD / Pkg of 1
COMPARE
21 3/4 1
$469.00USD / Pkg of 1
Showing 1- 10 of 10