Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore

Clear formulation for easier sample viewing
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Formulation: Tygon ND-100-80
Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.
Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.
Certifications: Resin meets USP Class VI requirements.
Temperature range: -25 to 185°F (-31 to 85°C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
$62.50 - $81.00USD / Roll of 1

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/100 1/32 1/100
$62.50USD / Roll of 1
COMPARE
1/50 1/16 1/50
$65.00USD / Roll of 1
COMPARE
1/32 3/32 1/32
$76.00USD / Roll of 1
COMPARE
1/25 5/64 1/64
$81.00USD / Roll of 1
COMPARE
1/20 3/32 1/50
$76.00USD / Roll of 1
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat