Masterflex® Transfer Tubing, Tygon® SE-200, Non-DEHP FEP-Lined

Keep samples free of odors and taste
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Formulation: Tygon® SE-200
Applications: Chemical and pharmaceutical processing, beverage dispensing, paint and solvent production, semiconductor processing
Characteristics: Bio-based, phthalate-free. Flexible, chemically resistant, inert, non-contaminating. Odorless, nontoxic, and taste free. Clear
Certifications: FDA compliant for contact food products
Temperature range: -49 to 165°F (-45 to 74°C)
Sterilize: Ethylene oxide
$116.00 - $1,541.00USD / Pkg of 1

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
85 1/8 1/4
$131.00USD / Pkg of 1
COMPARE
85 1/8 1/4
$1,541.00USD / Roll of 1
COMPARE
75 3/16 5/16
$202.00USD / Pkg of 1
COMPARE
55 1/4 3/8
$231.00USD / Pkg of 1
COMPARE
50 3/8 9/16
$316.00USD / Pkg of 1
COMPARE
45 1/2 3/4
$445.00USD / Pkg of 1
COMPARE
100 1/16 1/8
$116.00USD / Pkg of 1
COMPARE
30 3/4 1
$648.00USD / Pkg of 1
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat