Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3606
Pliable formulation for sharp radius bending
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
This tubing is ideal for general laboratory use. It offers long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, and is nontoxic.
FDA compliant and made according to GMP.
Temperature range is -51 to 165°F (-46 to 73°C)
Can be sterilized with ethylene oxide or autoclave
$33.10 - $1,603.00USD / Pkg of 1
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
36 1/16 1/8
$241.00USD / Case of 10
COMPARE
69 1/16 3/16
$85.70USD / Pkg of 1
COMPARE
47 3/32 7/32
$101.00USD / Pkg of 1
COMPARE
20 1/8 3/16
$63.40USD / Pkg of 1
COMPARE
36 1/8 1/4
$116.00USD / Pkg of 1
COMPARE
17 5/32 7/32
$71.20USD / Pkg of 1
COMPARE
25 3/16 5/16
$145.00USD / Pkg of 1
COMPARE
28 1/4 1/2
$361.00USD / Pkg of 1
COMPARE
23 5/16 1/2
$359.00USD / Pkg of 1
COMPARE
30 5/16 9/16
$308.00USD / Pkg of 1
COMPARE
14 3/8 1/2
$221.00USD / Pkg of 1
COMPARE
25 3/8 5/8
$427.00USD / Pkg of 1
COMPARE
13 7/16 9/16
$248.00USD / Pkg of 1
COMPARE
12 1/2 5/8
$249.00USD / Pkg of 1
COMPARE
20 1/2 3/4
$507.00USD / Pkg of 1
COMPARE
24 1/2 13/16
$582.00USD / Pkg of 1
COMPARE
13 5/8 13/16
$405.00USD / Pkg of 1
COMPARE
14 3/4 1
$550.00USD / Pkg of 1
COMPARE
12 1 1 1/4
$689.00USD / Pkg of 1
COMPARE
20 1 1 1/2
$1,603.00USD / Pkg of 1
Showing 1 - 20 of 30