Masterflex® UL-Approved Liquid Flow Switches for High Inline Pressures
Withstands pressures up to 1000 psi at 225°F (107°C)
  • UL-listed for use in Class I, Division 2, Groups A, B, C, D hazardous environments
These high-pressure SPDT switches withstand pressures up to 1000 psi at 225°F (107°C)! They provide reliable and consistent performance; repeatability is ±1%. Use a 50 µm filter to protect your switch (sold separately).
$253.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Wetted Materials Description
Availability Pricing
COMPARE
Brass, polysulfone, 316 stainless steel, Viton®, and epoxy UL-approved SPDT, 20VA Liquid Flow Switch for High Inline Pressures; 0.10 GPM
$253.00USD / Each
COMPARE
0.25 Brass, polysulfone, 316 stainless steel, Viton®, and epoxy UL-approved SPDT, 20 VA Liquid Flow Switch for High Inline Pressures; 0.25 GPM
$253.00USD / Each
COMPARE
0.5 Brass, polysulfone, 316 stainless steel, Viton®, and epoxy UL-approved SPDT 20 VA Liquid Flow Switch for High Inline Pressures; 0.50 GPM
$253.00USD / Each