Masterflex® Union Fittings, Hose Barb, Cross Connector
Quick and easy assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. Cross connectors feature four hose barb ports.
$17.40 - $96.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
COMPARE
5/32 Polypropylene Fitting, Polypropylene, Cross Connector, Hose Barb, 4 mm; 20/PK
$49.00USD / Pkg of 20
COMPARE
15/64 Polypropylene Fitting, Polypropylene, Cross Connector, Hose Barb, 6 mm; 20/PK
$73.00USD / Pkg of 20
COMPARE
5/16 Polypropylene Fitting, Polypropylene, Cross Connector, Hose Barb, 8 mm; 20/PK
$64.00USD / Pkg of 20
COMPARE
25/64 Polypropylene Fitting, Polypropylene, Cross Connector, Hose Barb, 10 mm; 20/PK
$70.00USD / Pkg of 20
COMPARE
15/32 Polypropylene Fitting, Polypropylene, Cross Connector, Hose Barb, 12 mm; 20/PK
$77.00USD / Pkg of 20
COMPARE
35/64 Polypropylene Fitting, Polypropylene, Cross Connector, Hose Barb, 14 mm; 20/PK
$88.00USD / Pkg of 20
COMPARE
1/4 PVDF Fitting, PVDF, Cross Connector, Hose Barb, 1/4"; 10/PK
$39.00USD / Pkg of 10
COMPARE
1/16 Stainless Steel Fitting, 316 Stainless Steel, Cross Connector, Hose Barb, 1/16"; EA
$67.50USD / Each
COMPARE
3/32 Stainless Steel Fitting, 316 Stainless Steel, Cross Connector, Hose Barb, 3/32"; EA
$61.50USD / Each
COMPARE
1/8 Stainless Steel Fitting, 316 Stainless Steel, Cross Connector, Hose Barb, 1/8"; EA
$62.50USD / Each
COMPARE
3/16 Stainless Steel Fitting, 316 Stainless Steel, Cross Connector, Hose Barb, 3/16"; EA
$75.00USD / Each
COMPARE
3/8 Stainless Steel Fitting, 316 Stainless Steel, Cross Connector, Hose Barb, 3/8"; EA
$96.00USD / Each
COMPARE
1/4 Polypropylene Fitting, Black Polypropylene, Cross Connector, Hose Barb, 1/4"; 5/PK
$20.50USD / Pkg of 5
COMPARE
1/4 Nylon Fitting, Nylon, Cross Connector, Hose Barb, 1/4"; 5/PK
$17.40USD / Pkg of 5
COMPARE
1/8 PVDF Fitting, PVDF, Cross Connector, Hose Barb, 1/8"; 10/PK
$38.00USD / Pkg of 10
Showing 1- 15 of 15