Masterflex® Union Fittings, Hose Barb, Straight, Nylon
Quick and easy assembly without tools
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
$15.00 - $152.00USD / Pkg of 100
35 variations of this product are available.
See All Product Options Below
35 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/16 3/16 Nylon
$20.75USD / Pkg of 10
COMPARE
3/8 3/8 Nylon
$20.25USD / Pkg of 10
COMPARE
1 1 Nylon
$24.75USD / Pkg of 10
COMPARE
1 1 Nylon
$24.00USD / Pkg of 10
COMPARE
1/4 1/4 Nylon
$19.50USD / Pkg of 10
COMPARE
5/16 5/16 Nylon
$18.70USD / Pkg of 10
COMPARE
3/8 3/8 Nylon
$19.50USD / Pkg of 10
COMPARE
1/2 1/2 Nylon
$19.50USD / Pkg of 10
COMPARE
5/32 5/32 Nylon
$22.50USD / Pkg of 10
COMPARE
3/16 3/16 Nylon
$15.80USD / Pkg of 10
COMPARE
5/8 5/8 Nylon
$90.50USD / Pkg of 100
COMPARE
1/4 1/4 Nylon
$86.00USD / Pkg of 100
COMPARE
3/8 3/8 Nylon
$83.50USD / Pkg of 100
COMPARE
5/32 5/32 Nylon
$111.00USD / Pkg of 100
COMPARE
3/16 3/16 Nylon
$111.00USD / Pkg of 100
COMPARE
1 1/4 1 1/4 Nylon
$22.75USD / Pkg of 5
COMPARE
5/32
$21.00USD / Pkg
COMPARE
1/4 Nylon
$17.40USD / Pkg
COMPARE
1/8 1/8 Nylon
$15.70USD / Pkg of 10
COMPARE
1/8 Black nylon
$15.00USD / Pkg
Showing 1 - 20 of 35