Masterflex® Union Fittings, Hose Barb, Straight, Stainless Steel
Quick and easy assembly without tools
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
$16.00 - $78.50USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
1/8 1/8 Stainless Steel
$16.00USD / Each
COMPARE
5/16 5/16 Stainless Steel
$24.75USD / Each
COMPARE
3/16 3/16 Stainless Steel
$19.90USD / Each
COMPARE
1/4 1/4 Stainless Steel
$36.00USD / Each
COMPARE
3/8 3/8 Stainless Steel
$38.00USD / Each
COMPARE
1/2 1/2 Stainless Steel
$67.50USD / Each
COMPARE
5/8 5/8 Stainless Steel
$72.00USD / Each
COMPARE
3/4 3/4 Stainless Steel
$78.50USD / Each
COMPARE
1/16 1/16 Stainless Steel
$51.00USD / Each
COMPARE
3/32 3/32 Stainless Steel
$63.50USD / Each
COMPARE
1/8 1/8 Stainless Steel
$48.75USD / Each
COMPARE
5/32 5/32 Stainless Steel
$60.75USD / Each
COMPARE
3/16 3/16 Stainless Steel
$51.00USD / Each
Showing 1- 13 of 13