Masterflex® Union Fittings, Hose Barb, Tee, Polypropylene
No tools needed for assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
$12.60 - $309.00USD / Pkg of 10
47 variations of this product are available.
See All Product Options Below
47 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
Polypropylene
$43.00USD / Pkg of 10
COMPARE
1/16 1/16 Polypropylene
$42.00USD / Pkg of 25
COMPARE
1/4 1/4 Polypropylene
$15.50USD / Pkg of 5
COMPARE
1/8 Polypropylene
$12.60USD / Pkg of 5
COMPARE
3/16 3/16 Polypropylene
$26.75USD / Pkg of 10
COMPARE
1/2 1/2 Polypropylene
$24.25USD / Pkg of 10
COMPARE
3/4 3/4 Polypropylene
$26.00USD / Pkg of 10
COMPARE
1/4 1/4 Polypropylene
$24.25USD / Pkg of 10
COMPARE
1 1 Polypropylene
$21.75USD / Pkg of 5
COMPARE
1/16 1/16 Polypropylene
$21.50USD / Pkg of 10
COMPARE
1/8 1/8 Polypropylene
$62.50USD / Pkg of 100
COMPARE
3/16 3/16 Polypropylene
$64.00USD / Pkg of 25
COMPARE
5/16 5/16 Polypropylene
$100.00USD / Pkg of 100
COMPARE
3/8 3/8 Polypropylene
$184.00USD / Pkg of 100
COMPARE
1/2 1/2 Polypropylene
$183.00USD / Pkg of 100
COMPARE
3/4 3/4 Polypropylene
$112.00USD / Pkg of 50
COMPARE
1 1 Polypropylene
$116.00USD / Pkg of 5
COMPARE
1/8 Polypropylene
$44.75USD / Pkg of 25
COMPARE
1/8 Polypropylene
$305.00USD / Pkg of 250
COMPARE
$309.00USD / Pkg of 250
Showing 1 - 20 of 47