Masterflex® Variable-Area Flowmeter Acrylic Tripod Bases
Easily mount flowmeters on tripod for easy viewing
  • Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
$58.25 - $141.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Material Housing Description
Availability Pricing
COMPARE
Acrylic Tripod Base for 03218-50 and 03218-52
$58.25USD / Each
COMPARE
Flowmeter Tripod Base for One Meter
$58.25USD / Each
COMPARE
Flowmeter Tripod Base for One, Two, or Three Meters
$67.25USD / Each
COMPARE
Acrylic Tripod Base for Three to Five Tube Systems
$58.25USD / Each
COMPARE
Acrylic Tripod Base for Multi-Tube Frames
$58.25USD / Each
COMPARE
Tripod base Flowmeter Stand For 32460
$137.00USD / Each
COMPARE
Tripod base Flowmeter Stand For 32461
$141.00USD / Each
Showing 1- 7 of 7