Masterflex® Variable Area Flowmeter Single Tube Frames
Create your own flowmeter systems for complete flexibility
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
  • Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
Choose a frame with a valve port if you plan on adding a valve cartridge to control flow.
$51.25 - $187.70USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Material Housing Valve Wetted Materials
Availability Pricing
COMPARE
Aluminum No Valve Port
$58.15USD / Each
COMPARE
Aluminum No Valve Port
$86.20USD / Each
COMPARE
Aluminum Valve Port
$51.25USD / Each
COMPARE
Aluminum Valve Port
$86.20USD / Each
COMPARE
Stainless Steel No Valve Port
$158.40USD / Each
COMPARE
Stainless Steel Valve Port
$158.40USD / Each
COMPARE
Stainless Steel Valve Port
$187.70USD / Each
Showing 1- 7 of 7