MECMESIN Multifunction Force Testing Systems

"Smart" tranducers let you expand the capabilities of your gauge – one meter works over multiple ranges
  • Multifunction gauges feature a high-speed 5000 Hz sampling rate, audio/visual alarms, and RS-232 interface
  • Display provides live readings and pass/fail leds allow quick test assessment
Measure tension, compression, or torque with these versatile, easy-to-use gauges. Choose a “smart” transducer to expand the capabilities of your gauge—meter automatically recognizes load cells and force/torque sensors. Optional foot switch controls up to two keypad functions for hands-free operation. Can be connected to the gauge or test stand.
$1,419.00 - $1,469.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs)
Availability Pricing
Mecmesin 850-416-V03 Multifunction Force Gauge; 11 lb EW-59885-12 Mfr # 850-416-V03
COMPARE
11
$1,469.00USD / Each
Mecmesin 850-418-V03 Multifunction Force Gauge; 55 lb EW-59885-13 Mfr # 850-418-V03
COMPARE
55
$1,459.00USD / Each
Mecmesin 850-419-V03 Multifunction Force Gauge; 110 lb EW-59885-14 Mfr # 850-419-V03
COMPARE
110
$1,458.50USD / Each
Mecmesin 850-420-V03 Multifunction Force Gauge; 220 lb EW-59885-15 Mfr # 850-420-V03
COMPARE
220
$1,419.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat