This product is now discontinued, View Related Products
MECMESIN Test Stands
Select a manual or single-column motorized test stand for accurate, repeatable test results. Single-column motorized test stands provide an accurate picture of how materials or products respond at any given point during compression or tension loading.
Stands feature crosshead speed from 0.04 to 40 in./min (1 to 1000 mm/min), single cycle test mode with full-speed crosshead return, auto-travel reverse on alarm point or sample break, headroom up to 20-3/4'' and presetable upper and lower travel limits.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Speed Range
Availability Pricing
Mecmesin 810-001 Single Column Motorized Test Stand, 115 VAC EW-59880-68 Mfr # 810-001
COMPARE
0.04 to 40 in/min (1 to 1000 mm/min)
Call For Price
Mecmesin 810-001 Single Column Motorized Test Stand, 230 VAC EW-59880-69 Mfr # 810-001
COMPARE
0.04 to 40 in/min (1 to 1000 mm/min)
Call For Price
Mecmesin 804-001 Test Stand for 59885-Series Force Gauges; 220lb/100kg/1000N EW-59880-71 Mfr # 804-001
COMPARE
Manual