MegaMag Genie Magnetic Stirrers
Ideal for your large-capacity stirring needs
  • Powerful magnet and motor capable of stirring up to 38 L
Stirrers feature a powerful heavy-duty motor, making them ideal for all of your large-capacity stirring needs. The spill-resistant, durable ABS/polycarbonate housing is designed to channel fluids away from internal components. Internal ramping feature will gradually increase speed to the set point to minimize splashing and improve control of the stir bar. Choose between the analogue dial or digital true-speed control. Digital version features a LED display for easy speed indication and can be controlled in 10-rpm increments. Digital stirrer can be set to run continuously or for a timed run from 1 to 99 minutes.
$887.00 - $1,195.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Power (VAC) Description
Availability Pricing
Scientific Industries Genie SI-2236 MegaMag , Large-Capacity Analog Magnetic Stirrer; 120V EW-86575-10 Mfr # SI-2236
COMPARE
1300 120 MegaMag , Large-Capacity Analog Magnetic Stirrer; 120V
$887.00USD / Each
Scientific Industries Genie SI-2256 MegaMag , Large-Capacity Analog Magnetic Stirrer; 230V EW-86575-12 Mfr # SI-2256
COMPARE
1300 230 MegaMag , Large-Capacity Analog Magnetic Stirrer; 230V
$965.00USD / Each
Scientific Industries Genie SI-3236 Digital MegaMag Stirrer, 120 VAC EW-86575-15 Mfr # SI-3236
COMPARE
1300 120 Digital MegaMag Stirrer, 120 VAC
$1,180.00USD / Each
Scientific Industries Genie SI-3236L Digital MegaMag Low Range Stirrer, 120 VAC EW-86575-19 Mfr # SI-3236L
COMPARE
300 120 Digital MegaMag Low Range Stirrer, 120 VAC
$1,195.00USD / Each