This product is now discontinued, View Related Products
Megger 247000 Series Digital Low Resistance Ohmmeters
Four wire measurement eliminates lead and contact resistances
  • Automatic zero circuit eliminates field test accuracies
  • Broad line of test leads simplifies data collection
  • Standard accuracy of ±0.25%
These rugged, portable ohmmeters provided accuracies of ±0.25% and resolution down to 0.1 µΩ. The ohmmeters are simple to operate, just connect the leads, turn the unit on, and read the measurement directly from the digital LED display. No warm-up is required. Applications include measuring the resistance of switch and circuit breaker contacts, transformers and generators, motor windings, busbar joints, aircraft, rail and pipe bonds, cable splices, metal alloys, graphite electrodes, and wire and cable.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
4-Wire Resistance Max Resistance (M ohms) Resistance Accuracy
Availability Pricing
Megger 247000 Digital Low Resistance Ohmmeter, Dual-Pak; 10 Amp EW-20023-03 Mfr # 247000
COMPARE
Yes 5.999 ±0.25% of reading = 1 LSD
Call For Price
Megger 247001 Digital Low Resistance Ohmmeter, Single-Pak; 10 Amp EW-20023-04 Mfr # 247001
COMPARE
Yes 5.999 ±0.25% of reading = 1 LSD
Call For Price
Megger 247002 Digital Low Resistance Ohmmeter, Single-Pak Low Range; 10 Amp EW-20023-05 Mfr # 247002
COMPARE
Yes 5.999 0.3% of reading = 1 LSD