Megger Digital Hydrometer Kits
Provides memory for both temperature and specific gravity for up to eight tests
  • Measure electrolyte specific gravity (SG) and temperature
  • 5-second measurements
  • Save up to 8 tests in the 8-channel data logger
  • Import the specific gravity data directly into Megger Power DB
The digital hydrometer kit is an eight-channel digital hydrometer kit that accurately and quickly determines specific gravity of flooded cells. Simply draw the electrolyte into it and the specific gravity and temperature will be determined in five seconds. It provides memory for both temperature and specific gravity for eight tests of 256 cells each. The stored data can be easily transferred to the accompanying data logger which can hold 8 separate sets of test data. This data can then be easily imported directly into a Megger Power DB battery report from the data logger.
$3,180.00 - $10,325.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Density Range (g/cm³) Description
Availability Pricing
Megger Eight Channel Digital Hydrometer Kit EW-20027-05 Mfr # 2001-692
COMPARE
3 Eight Channel Digital Hydrometer Kit
$10,325.00USD / Each
Megger Hydrometer Data Logger Kit for Anton Paar Hydrometers EW-20027-07 Mfr # 2001-693
COMPARE
3 Hydrometer Data Logger Kit for Anton Paar Hydrometers
$3,180.00USD / Each