This product is now discontinued, View Related Products

Megger Digital Low Resistance Microhmmeters

Results in under 3 seconds with 0.2% basic accuracy
  • Water resistant to IP65
  • Auto current reversal cancels standing emfs
  • Fuse protected to 600V with 250 mW power limit (with optional override) to avoid heating the test sample
  • Automatically detects continuity in potential and current connections
  • Provides visible warning of voltages present at the terminals
  • Multiple operating modes including fully automatic
These digital low resistance microhmmeters provide accurate results in under three seconds. Select suitable test current, up to 10 A dc to measure reistance from 0.1 µΩ to 2000 Ω on one of sevent ranges. Consturcted with a strong, lightweight case, the units are well suited for use in the field or in the laboratory.
3 PRODUCT OPTIONS
Compareitem specifications
AC Current Range (mA) Max Resistance (ohms) Resistance Resolution
Availability Pricing
Megger 249003 CALIBRTN SHUNT 0.01OHMS 10A EW-20011-18 Mfr # 249003
COMPARE
Call For Price
Megger DLRO10 Digital Low Resistance Microhmmeter; 10 Amp EW-20022-29 Mfr # 1006-598
COMPARE
±10% 1999.9 0.1 μΩ;1 μΩ;10 μΩ;100 μΩ;1 mΩ;10 mΩ;100 mΩ
Call For Price
Megger DLRO10X Digital Low Resistance Microhmmeter; 10 Amp EW-20022-30 Mfr # 1006-600; plus DH4-C
COMPARE
±10% 1999.9 0.1 μΩ;1 μΩ;10 μΩ;100 μΩ;1 mΩ;10 mΩ;100 mΩ
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat