Megger Piezo Verifier

Safely confirm operation of voltage and phasing testers up to 30 kV
  • Small compact design easily allows the checking of the integrity of current paths and operation of indicators
  • Safely ensure IEC conformance
This essential safety tool is used to confirm the correct operation of voltage detectors and phasing testers that do not have a self-testing capability. When the verifier handle is depressed at the rate of 3 strokes per second, a piezo crystal generates a safety check voltage. This voltage, when applied to the tip of the detector, tests for integrity of the current path and proper operation of the indicators.

A three step proedure of testing the detector with the piezo verifier, using the detector to test for the absence of high voltage and then testing the detector again should be strictly observed in accordance with accepted operating procedures. Verifier can be used with single and two pole models.
$635.00 - $660.00USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Voltage Range (Voltage)
Availability Pricing
Megger 510005 Piezo Verifier, , generates 1800 V EW-20022-68 Mfr # 510005
COMPARE
1800
$635.00USD / Each
Megger 510006 Piezo Verifier, generates 10, 20 and 30 kV EW-20022-69 Mfr # 510006
COMPARE
30000
$660.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat