Meiji Inverted Microscopes
Inverted design offers extra working distance and increases allowable sample size
  • Infinity-corrected optics provide crisp, distortion free, high-resolution images
Meiji inverted microscopes are rugged and ergonomically designed for long-lasting and comfortable use. The inverted design accommodates a wide variety of glass and plastic vessels and flasks as well as larger sample sizes.
An inclined and adjustable siedentopf type viewing head provides ergonomic viewing. The nosepiece is quintuple turret with precision click stops. Microscopes feature a variable rheostat, automatic voltage sensing power supply, and a 247 mm x 180 mm plain mechanical stage with low-position coaxial coarse and fine-focus control knobs. A front-mounted camera port accommodates 35 mm CCD, and digital photomicrographic attachments.
$5,390.00 - $6,040.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Illuminator(s) Maximum Magnification Power (VAC)
Availability Pricing
Meiji Techno Inverted Microscope, Brightfield, 115 VAC EW-48405-02 Mfr # TC-5100
COMPARE
30 W halogen Koehler 200x 115
$5,390.00USD / Each
Meiji Techno Inverted Microscope, Phase contrast, 115 VAC EW-48405-06 Mfr # TC-5300
COMPARE
30 W halogen Koehler with phase slider 200x 115
$6,040.00USD / Each
Meiji Techno Inverted Microscope, Phase Contrast, 220 VAC EW-48405-08 Mfr # TC-5300/220
COMPARE
30 W halogen Koehler with phase slider 200x 220
$6,040.00USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat