Meiji Techno Microscope Accessories

Optimize life and performance of your Meiji Techno microscopes
  • Keep your Meiji Techno microscopes running smoothly
Choose from a wide range of accessories and replacement parts to enhance performance and optimize the life of your Meiji Techno microscopes.
$13.40 - $79.80USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
Meiji Techno Microscopy Immersion Oil, 7.4 mL EW-48401-95 Mfr # MA700
COMPARE
Oil immersion microscope objectives Microscopy Immersion Oil, 7.4 mL
$13.40USD / Each
Meiji Techno Replacement Microscope Eyeshields, Rubber; Pair EW-48402-65 Mfr # MA600
COMPARE
Meiji Techno EMZ Series, EMF Series, EMT Series eyepieces Replacement Microscope Eyeshields, Rubber; Pair
$25.80USD / Pair of 1
Meiji Techno Replacement Microscope Stage Clips, 75 mm L x 4 mm; Pair EW-48404-70 Mfr # MA593
COMPARE
Meiji Techno microscopes Replacement Microscope Stage Clips, 75 mm L x 4 mm; Pair
$30.70USD / Pair of 2
Meiji Techno Microscope Camera Adapter Ring, Canon EOS EW-48407-82 Mfr # T2-9
COMPARE
Canon EOS camera Microscope Camera Adapter Ring, Canon EOS
$79.80USD / Each
Meiji Techno Microscope Camera Adapter Ring, Nikon EW-48407-83 Mfr # T2-5
COMPARE
Nikon camera Microscope Camera Adapter Ring, Nikon
$79.80USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat