Meriam Digital Manometer and Single Function Calibrators
  • Lightweight, ergonomic, user friendly
  • 5 digit LCD display with 0.4'' high digits
  • Record mode lets user capture up to 240 readings
  • Digital zero and span adjustment
  • 8 selectable user units
  • Ranges from 28 inH2O to 100 PSI
  • 3xAA battery, field replaceable with >100 hours continuous use
  • Auto power off after 20 minutes
  • Media compatibility; non isolated sensors for clean, dry, non-corrosive gases
  • Low battery indicator
  • Process connection 1/8'' female NPT
  • Protective rubber boot
  • Horizontal bar graph for full scale pressure reference
  • Accuracy ± .50 fso standard
$525.00 - $607.00USD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Meriam M101-GN005 Digital Manometer and Calibrator, 0-5 psig EW-10271-46 Mfr # M101-GN005
COMPARE
±0.1% full-scale 5
$607.00USD / Each
Meriam M100-GN0015 Digital Manometer, 0-15 psi Gauge EW-10323-16 Mfr # M100-GN0015
COMPARE
±0.25% full-scale 15
$525.00USD / Each
Meriam M100-GN0050 Digital Manometer, 0-50 psi Gauge EW-10323-17 Mfr # M100-GN0050
COMPARE
±0.25% full-scale 50
$527.00USD / Each
Meriam M100-DN0832 Digital Manometer, 0-832"WC Differential EW-10323-18 Mfr # M100-DN0832
COMPARE
±0.25% full-scale
$525.00USD / Each
Meriam M100-AN1551 Digital Manometer, 0-1551 mmHg Absolute EW-10323-19 Mfr # M100-AN1551
COMPARE
±0.25% full-scale
$525.00USD / Each
Meriam M101-DN0138 Digital Manometer/Calibrator, 0-138" WC Differential EW-10323-22 Mfr # M101-DN0138
COMPARE
±0.1% full-scale
$607.00USD / Each
Meriam M101-DN0832 Digital Manometer/Calibrator, 0-832" WC Differential EW-10323-23 Mfr # M101-DN0832
COMPARE
±0.1% full-scale
$607.00USD / Each
Meriam M101-AN1551 Digital Manometer/Calibrator, 0-1551 mmHg Absolute EW-10323-24 Mfr # M101-AN1551
COMPARE
±0.1% full-scale
$607.00USD / Each
Meriam M100-GN005 Digital Manometer, 0-5 psi Gauge EW-10323-37 Mfr # M100-GN005
COMPARE
±0.25% full-scale 5
$525.00USD / Each
Meriam M100-DN0138 Digital Manometer, 0-138"WC Differential EW-25751-11 Mfr # M100-DN0138
COMPARE
±0.25% full-scale
$525.00USD / Each
Meriam M100-DN0028 Digital Manometer, 0-28" WC Differential EW-68460-30 Mfr # M100-DN0028
COMPARE
±0.25% full-scale
$530.00USD / Each
Meriam M100-GN0100 Digital Manometer, 0-100 psi Gauge EW-68460-32 Mfr # M100-GN0100
COMPARE
±0.25% full-scale 100
$525.00USD / Each
Meriam M100-DN0416 Digital Manometer, 0-416" WC Differential EW-68460-33 Mfr # M100-DN0416
COMPARE
±0.25% full-scale
$525.00USD / Each
Meriam M100-AN2585 Digital Manometer, 0-2585 mmHg Absolute EW-68460-34 Mfr # M100-AN2585
COMPARE
±0.25% full-scale
$525.00USD / Each
Meriam M101-DN0028 Digital Manometer/Calibrator, 0-28" WC Differential EW-68460-35 Mfr # M101-DN0028
COMPARE
±0.1% full-scale
$607.00USD / Each
Meriam M101-GN0030 Digital Manometer/Calibrator, 0-30 psi Gauge EW-68460-36 Mfr # M101-GN0030
COMPARE
±0.1% full-scale 30
$607.00USD / Each
Meriam M101-GN0100 Digital Manometer/Calibrator, 0-100 psi Gauge EW-68460-37 Mfr # M101-GN0100
COMPARE
±0.1% full-scale 100
$607.00USD / Each
Meriam M101-AN2585 Digital Manometer/Calibrator, 0-2585 mmHg Absolute EW-68460-39 Mfr # M101-AN2585
COMPARE
±0.1% full-scale
$607.00USD / Each
Meriam M101-AN0775 Digital Manometer/Calibrator, 0-775 mmHg Absolute EW-68863-72 Mfr # M101-AN0775
COMPARE
±0.1% full-scale
$607.00USD / Each
Meriam M100-AN0775 Digital Manometer, 0-775 mmHg Absolute EW-68874-11 Mfr # M100-AN0775
COMPARE
±0.25% full-scale
$525.00USD / Each
Showing 1- 20 of 20