Metal Band Drum Heaters

Achieve better flow of viscous liquids
These electric heaters combine adjustable temperature control with a variable power output selector. This output selector provides three wattage levels for a given thermostat setting and voltage rating. A pilot light indicates when the heating element is activated. Temperature detector is located on the outside of the pail or drum away from the active heating element. Caution: Use heaters with metal pails or drums that contain nonflammable materials only. Also, heaters are only for indoor use in nonhazardous areas.
$672.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Max Temperature (° F) Power (VAC)
Availability Pricing
Expo Engineering TRX-55-L/R-115V Drum Heater, 55-Gallon, 60 to 250°F, 2000 Watts, 120 V EW-09399-50 Mfr # TRX-55-L/R-115V
COMPARE
55 gallons 250 120
$672.00USD / Each
Expo Engineering TRX-55-L/R-220V Drum Heater, 55-Gallon, 60 to 250°F, 3000 Watts, 240 V EW-09399-52 Mfr # TRX-55-L/R-220V
COMPARE
55 gallons 250 240
$672.00USD / Each
Expo Engineering TRX-55-H/R-115V Drum Heater, 55-Gallon, 200 to 400°F, 2000 Watts, 120 V EW-09399-53 Mfr # TRX-55-H/R-115V
COMPARE
55 gallons 400 120
$672.00USD / Each
Expo Engineering TRX-55-H/R-220V Drum Heater, 55-Gallon, 200 to 400°F, 3000 Watts, 240 V EW-09399-54 Mfr # TRX-55-H/R-220V
COMPARE
55 gallons 400 240
$672.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat