Metal Industrial Ball Valves
  • Full flow design ensures maximum operating efficiency
  • Blow-out proof stems
Speically-designed handle increases turning leverage and provides easy opening and closing. Brass valves are an ideal choice for use in water and air service lines. Stainless steel valves are ideal for use in corrosive environments such as chemical plants, refineries, and steel mills.
$28.35 - $353.60USD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Outlet Port
Availability Pricing
Parker Hannifin XV500P-4 1/4" NPT brass female/female manual ball valves EW-30527-00 Mfr # XV500P-4
COMPARE
4.0 1/4" NPT(F) 1/4" NPT(F)
$29.55USD / Each
Parker Hannifin XV500P-6 3/8" NPT brass female/female manual ball valves EW-30527-02 Mfr # XV500P-6
COMPARE
5.8 3/8" NPT(F) 3/8" NPT(F)
$28.35USD / Each
Parker Hannifin XV500P-8 1/2" NPT brass female/female manual ball valves EW-30527-04 Mfr # XV500P-8
COMPARE
12.0 1/2" NPT(F) 1/2" NPT(F)
$28.35USD / Each
Parker Hannifin XV500P-12 3/4" NPT brass female/female manual ball valves EW-30527-06 Mfr # XV500P-12
COMPARE
25.0 3/4" NPT(F) 3/4" NPT(F)
$46.55USD / Each
Parker Hannifin XV500P-16 1" NPT brass female/female manual ball valves EW-30527-08 Mfr # XV500P-16
COMPARE
35.0 1" NPT(F) 1" NPT(F)
$58.50USD / Each
Parker Hannifin XV501P-4 1/4" NPT brass female/male manual ball valves EW-30527-20 Mfr # XV501P-4
COMPARE
6.3 1/4" NPT(F) 1/4" NPT(M)
$39.50USD / Each
Parker Hannifin XV501P-6 3/8" NPT brass female/male manual ball valves EW-30527-22 Mfr # XV501P-6
COMPARE
5.7 3/8" NPT(F) 3/8" NPT(M)
$42.15USD / Each
Parker Hannifin XV501P-8 1/2" NPT brass female/male manual ball valves EW-30527-24 Mfr # XV501P-8
COMPARE
10.0 1/2" NPT(F) 1/2" NPT(M)
$40.05USD / Each
Parker Hannifin XV501P-12 3/4" NPT brass female/male manual ball valves EW-30527-26 Mfr # XV501P-12
COMPARE
25.0 3/4" NPT(F) 3/4" NPT(M)
$55.65USD / Each
Parker Hannifin XV501P-16 1" NPT brass female/male manual ball valves EW-30527-28 Mfr # XV501P-16
COMPARE
35.0 1" NPT(F) 1" NPT(M)
$82.10USD / Each
Parker Hannifin XV502SS-4 1/4" NPT stainless steel female/female manual ball valve EW-30528-00 Mfr # XV502SS-4
COMPARE
4.0 1/4" NPT(F) 1/4" NPT(F)
$108.20USD / Each
Parker Hannifin XV502SS-6 3/8" NPT stainless steel female/female manual ball valve EW-30528-02 Mfr # XV502SS-6
COMPARE
6.0 3/8" NPT(F) 3/8" NPT(F)
$106.10USD / Each
Parker Hannifin XV502SS-8 1/2" NPT stainless steel female/female manual ball valve EW-30528-04 Mfr # XV502SS-8
COMPARE
14.0 1/2" NPT(F) 1/2" NPT(F)
$108.20USD / Each
Parker Hannifin XV502SS-12 3/4" NPT stainless steel female/female manual ball valve EW-30528-06 Mfr # XV502SS-12
COMPARE
35.0 3/4" NPT(F) 3/4" NPT(F)
$188.30USD / Each
Parker Hannifin XV502SS-16 1" NPT stainless steel female/female manual ball valve EW-30528-08 Mfr # XV502SS-16
COMPARE
54.0 1" NPT(F) 1" NPT(F)
$253.60USD / Each
Parker Hannifin XV501SS-4 1/4" NPT stainless steel female/male manual ball valve EW-30529-00 Mfr # XV501SS-4
COMPARE
4.0 1/4" NPT(F) 1/4" NPT(M)
$146.60USD / Each
Parker Hannifin XV501SS-6 3/8" NPT stainless steel female/male manual ball valve EW-30529-02 Mfr # XV501SS-6
COMPARE
6.0 3/8" NPT(F) 3/8" NPT(M)
$155.00USD / Each
Parker Hannifin XV501SS-8 1/2" NPT stainless steel female/male manual ball valve EW-30529-04 Mfr # XV501SS-8
COMPARE
14.0 1/2" NPT(F) 1/2" NPT(M)
$152.90USD / Each
Parker Hannifin XV501SS-12 3/4" NPT stainless steel female/male manual ball valve EW-30529-06 Mfr # XV501SS-12
COMPARE
35.0 3/4" NPT(F) 3/4" NPT(M)
$224.70USD / Each
Parker Hannifin XV501SS-16 1" NPT stainless steel female/male manual ball valve EW-30529-08 Mfr # XV501SS-16
COMPARE
54.0 1" NPT(F) 1" NPT(M)
$353.60USD / Each
Showing 1- 20 of 20