Metal Tee Fittings 49372
Ensure proper insertion depth and measurement
These installation fittings are suitable for use with our paddle wheel flow sensor systems. Complete systems require slow sensor, installation fitting, and monitor or controller. Other components sold separatley.
The carbon steel installation fittings weld directly onto hole cut in pipe, and are ideal for stainless steel and carbon steel pipes.
The copper installation fitting is suitable for copper, 316 stainless steel (SS), carbon steel (CS), and galvanized iron (schedule 40) pipes. Copper tee fittings include PVDF insert for pipe ID over 1" and feature sweat-on ends.
$475.00 - $563.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Pipe Size Wetted Materials
Availability Pricing
GF Signet CS4T005 Carbon Steel Tee 0.5" EW-05620-51 Mfr # CS4T005
COMPARE
$475.00USD / Each
GF Signet CS4T007 Carbon Steel Tee 0.75" EW-05620-52 Mfr # CS4T007
COMPARE
$475.00USD / Each
GF Signet CUKT020 Copper Tee Fitting 2in Pipe Id EW-05620-76 Mfr # CUKT020
COMPARE
$563.00USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat