Metalex Paddle-Wheel Flow Sensors
Stainless steel body withstands high pressures and temperatures
  • Use in agressive liquid flow applications
Use these rugged sensors to monitor the flow of liquid ammonia, turbine steam condensate, and more—wherever strength and chemical resistance are required.
Open-core rotor is cavitation free at flow rates from 1.5 to 20 ft/second. Easy maintenance—sensors can be replaced in minutes.
METALEX flow sensors are compatible with all of our battery- and line-powered flowmeters, accumulators, and controllers. Sensors must be used with the fitting type specified in the table below—installation fittings sold separately.
METALEX sensors feature CD4MCU rotor; Fluoroloy B rotor bearings; and 316 stainless steel body, rotor shaft, and shaft retainers.
$901.00 - $929.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (")
Availability Pricing
GF Signet P525-1 Metalex Paddle-Wheel Flow Sensors, for use with mini-tap fitting; 1-1/2" EW-05618-60 Mfr # P525-1
COMPARE
1 1/2
$901.00USD / Each
GF Signet P525-2 Metalex Paddle-Wheel Flow Sensors, for use with mini-tap fitting; 2-1/2" EW-05618-64 Mfr # P525-2
COMPARE
2 1/2
$929.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat