Meterman LCR - CR Meters
Available for sale only in the United States and Canada
The MNCR50 is a pocket-sized capacitance and resistance meter that measures capacitance in 9 ranges from 200pF to 20mF; seven ranges for resistance from 20 ohm to 20Mohm. The MNLCR55 measures inductance from 200uH to 200H; capacitance measures ranges from 200pF to 20mF, and resistance from 20ohm to 20Mohm.
ITEM(S) ON SALE
$354.59USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Resistance Accuracy Resistance Range Resistance Resolution
Availability Pricing
Amprobe LCR55A LCR Tester with Transistor Test EW-20021-45 Mfr # LCR55A
COMPARE
0.01 ohm in 20 ohm 20M ohm
$354.59USD / Each
$393.99USD / Each