This product is now discontinued, View Related Products

Fisher Chemical Methyl α-D-Mannopyranoside

  • Assay - ≥99%
  • FTIR - Conforms to standard
  • Melting Point - 187-197°C
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical BP253010 Methyl alpha-D-mannopyranoside (10g) EW-88060-47 Mfr # BP253010
COMPARE
617-04-9
Call For Price
Fisher Chemical BP2530100 Methyl alpha-D-mannopyranoside (100g) EW-88060-48 Mfr # BP2530100
COMPARE
617-04-9
Call For Price
Fisher Chemical BP253025 Methyl alpha-D-mannopyranoside (25g) EW-88060-49 Mfr # BP253025
COMPARE
617-04-9
Call For Price
Fisher Chemical BP2530500 Methyl alpha-D-mannopyranoside (500g) EW-88060-50 Mfr # BP2530500
COMPARE
617-04-9
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat