Acros Organics Methyl Orange
Ideal for used in titration for acids
  • Shows red color in acidic medium and yellow color in basic medium
Methyl orange is a pH indicator frequently used in titration because of its clear and distinct color variance at different pH values. Because it changes color at the pKa of a mid strength acid, it is usually used in titration for acids.
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$81.60 - $160.25USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (g)
Availability Pricing
Acros Organics Methyl Orange, indicator (100g) CAS 547-58-0 EW-88153-50 Mfr # AC151421000
COMPARE
547-58-0 Pure 100
$82.80USD / Each
Acros Organics Methyl Orange, reagent ACS (25g) CAS 547-58-0 EW-88351-70 Mfr # AC414660250
COMPARE
547-58-0 ACS Reagent 25
$81.60USD / Each
Acros Organics Methyl Orange, 0.1% w/v solution (500ml) EW-88365-05 Mfr # AC611285000
COMPARE
547-58-0 500
$160.25USD / Each