This product is now discontinued, View Related Products

Acros Organics Methylacrylate, Stabilized, 99%

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC126190010 Methylacrylate, stabilized 99% (1l) EW-88125-99 Mfr # AC126190010
COMPARE
96-33-3
Call For Price
Acros Organics AC126190025 Methylacrylate, stabilized 99% (2.5l) EW-88126-00 Mfr # AC126190025
COMPARE
96-33-3
Call For Price
Acros Organics AC126190100 Methylacrylate, stabilized 99% (10ml) EW-88126-01 Mfr # AC126190100
COMPARE
96-33-3
Call For Price
Acros Organics Methylacrylate, stabilized 99% (500ml) EW-88126-02 Mfr # AC126195000
COMPARE
96-33-3
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat