Mettler Toledo Compact Volumetric Karl Fischer Titrators
Compact yet full of features to provide accurate water content determination
  • Unique cell design ensures accurate and repeatable measurements
  • Easy-to-understand One Click® touchscreen user interface
  • Automatic sensor and burette recognition
  • Burette with RFID chip saves all relevant titrant data
  • Solvent manager provides safe chemical handling
  • Flexible data export via two USB ports
These compact volumetric Karl Fischer titrators offer everything you need for fast and precise water content determination in an easy-to-use system. Analyze samples with water content between 100 ppm to 100% in single measurements or sample series with statistics. Robust design and easy-to-use interface make these simple to use.
Special titration cell design ensures accurate and repeatable measurements by minimizing ambient moisture interference, resulting in a low background drift value. Use the large color touchscreen to operate the simple-to-use One Click user interface to customize home screens to run your workflows with a single tap. Each user can set their own home screen with customized language selection (English, German, French, Spanish, Italian, Chinese, Russian, Polish, Korean, or Portuguese) and shortcut keys.
Automatic sensor and burette recognition reduces extra data entry steps and helps minimize errors. Burettes feature an RFID chip that contains concentration or lifetime of the titrant and is directly transferred to the titrator. Ensure user safety with the solvent manager. It is fully controlled by the titrator, and offers filling, draining, and solvent exchange without additional user interaction. For secure titrator operation, designate user groups to allow full or limited access. Easily export data in CSV, PDF, or XML files using a memory stick or connect to a printer via the USB port.
$10,500.00 - $12,500.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Output Power (VAC) Description
Availability Pricing
Mettler Toledo V10S Compact Volumetric Karl Fischer Titrator with 4 Shortcuts EW-78907-26 Mfr # 30267116
COMPARE
USB (2), Ethernet 100 to 240 V10S Compact Volumetric Karl Fischer Titrator with 4 Shortcuts
$10,500.00USD / Each
Mettler Toledo V20S Compact Volumetric Karl Fischer Titrator with 12 Shortcuts EW-78907-27 Mfr # 30252666
COMPARE
USB (2), Ethernet 100 to 240 V20S Compact Volumetric Karl Fischer Titrator with 12 Shortcuts
$12,500.00USD / Each