Mettler Toledo Deionizer 64439
No more scattering of powdery samples
  • Electrode removes electrostatic discharge reducing contamination and increasing productivity
  • Place electrode next to balance for best results
This deionizer dissipates static charge from the surrounding area, including your sample. Get accurate, reliable results at a quicker pace. Available with or without a warning light.
ITEM(S) ON SALE
$1,788.15 - $2,302.30USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo 63052302 Haug Deionizer, with Warning Light EW-11336-95 Mfr # 63052302
COMPARE
Haug Deionizer, with Warning Light
$2,302.30USD / Each
$2,530.00USD / Each
Mettler Toledo 63052303 Haug Deionizer, without Warning Light
Always in Stock
EW-11336-96 Mfr # 63052303
COMPARE
Haug Deionizer, without Warning Light
$1,788.15USD / Each
$1,965.00USD / Each