Mettler Toledo LiquiPhysics™ Excellence Density Meters
Efficiently and accurately measure the specific gravity of practically any type of sample
  • Fast results with a measuring time of only 30 seconds
  • Small footprint allows for use where bench space is limited
  • Integrated Peltier thermostat for excellent temperature control
These density meters are easy to use with a high level of accuracy and reliability. These units feature quick measurements including a unique method/product concept with automatic error detection and automatic adjustment verifications. All models offer extremely fast heating up- and cooling down rates with temperature control and will provide reliable results even prior to thermal equilibrium being reached. The built-in algorithm shortens the time required per measurement by 75% as well as detects non-homogeneous measuring cell filling.
The models differ only in their measuring accuracy. The DM45 DeltaRange and DM50 models have a five decimal place measuring accuracy, which is rarely needed over the entire density and temperature measuring ranges for density determinations. Only samples such as organic solvents and alcohol/water mixtures sometimes need to be measured with five decimal place accuracy. All these products have densities lower than 1 g/cm3 and the measurements are normally carried out at temperatures between 15 and 20°C. The DeltaRange density measuring cells are tailor-made solutions for such applications as they offer five decimal place accuracy in exactly this range at a lower cost than full range five decimal place cells.
$14,370.00 - $25,330.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Density Range (g/cm³) Description
Availability Pricing
Mettler Toledo DM40 LiquiPhysics Excellence Density Meter, 0.0001 g/cm3 EW-33333-34 Mfr # 51337000
COMPARE
3 Mettler Toledo DM40 LiquiPhysics Excellence Density Meter, 0.0001 g/cm3
$14,370.00USD / Each
Mettler Toledo DM45 LiquiPhysics Excellence Density Meter, 0.00005 g/cm3 EW-33333-35 Mfr # 51337001
COMPARE
3 Mettler Toledo DM45 LiquiPhysics Excellence Density Meter, 0.00005 g/cm3
$17,520.00USD / Each
Mettler Toledo DM50 LiquiPhysics Excellence Density Meter, 0.00002 g/cm3 EW-33333-36 Mfr # 51337002
COMPARE
3 Mettler Toledo DM50 LiquiPhysics Excellence Density Meter, 0.00002 g/cm3
$25,330.00USD / Each