Mettler Toledo NewClassic ME-T Analytical Balances
Easy worry-free weighing
  • Large color touchscreen and intuitive interface simplify operation
  • Fully Automatic Calibration Technology (FACT) ensures accuracy
  • Eight embedded applications include essential weighing functions, statistics, and formulation
  • Sample ID and passcode protection produce GLP documentation of results
  • Built-in connectivity includes RS-232, USB host, and USB device
ANSI connectors are white
ITEM(S) ON SALE
$3,508.05 - $4,199.65USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo ME54TE NewClassic ME-TE  Analytical Balance, 52 g x 0.1 mg EW-11335-59 Mfr # ME54TE
COMPARE
External 52 0.0001
$3,508.05USD / Each
$3,855.00USD / Each
Mettler Toledo ME104TE NewClassic ME-TE Analytical Balance, 120 g x 0.1 mg EW-11335-60 Mfr # ME104TE
COMPARE
External 120 0.0001
$4,160.00USD / Each
Mettler Toledo ME204TE NewClassic ME-TE Analytical Balance, 220 g x 0.1 mg EW-11335-61 Mfr # ME204TE
COMPARE
External 220 0.0001
$4,199.65USD / Each
$4,615.00USD / Each