Mettler Toledo NewClassic MS Toploading Balances
Fast settling time and consistent results for routine to complicated weighing procedures
  • Fully automatic time- and temperature-controlled internal calibration adjustment (FACT)
  • Advanced MonoBloc weighing technology provides fast and accurate results
  • Durable high-grade chemical-resistant die-cast aluminum housing
  • Overload Protection guards the weighing cell against excess weight overload
  • SmartTrac software displays how much of the entire weighing range has been used
  • Direct connection to a PC or printer via RS232 or USB slot allows convenient transfer of weighing results
  • High-contrast display (HCD) and intuitive menu for simple operation
  • Programmable Smart Keys for shortcut access to preferred applications
  • IP65 rating when used with the included in-use cover
These balances feature a robust housing made of high quality chemical-resistant aluminum that protects the sensitive sensor from environmental influences and impacts. The proven MonoBloc weighing technology and FACT – fully automatic time- and temperature-controlled internal adjustment – provide accurate, repeatable results for everyday weighing tasks. A new High Contrast Display with big numbers, clear symbols and an intuitive menu with new programmable SmartKeys allows users to operate the balance quickly and easily with one touch operation.
$6,280.00 - $8,820.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Mettler Toledo MS12001L Newclassic MS Toploading Balance, 12200g x 0.1g EW-11334-71 Mfr # MS12001L
COMPARE
12200 115 0.1
$6,280.00USD / Each
Mettler Toledo MS16001L Newclassic MS Toploading Balance, 16200g x 0.1g EW-11334-72 Mfr # MS16001L
COMPARE
16200 115 0.1
$7,130.00USD / Each
Mettler Toledo MS32001L Newclassic MS Toploading Balance, 32200g x 0.1g EW-11334-73 Mfr # MS32001L
COMPARE
32200 115 0.1
$8,820.00USD / Each
Mettler Toledo MS32000L Newclassic MS Toploading Balance, 32200g x 1g EW-11334-74 Mfr # MS32000L
COMPARE
32200 115 1
$6,340.00USD / Each