METTLER TOLEDO SevenExcellence™ Benchtop pH/Conductivity Meters
Easy to set up and view all measurements simultaneously
  • Intuitive plain-language interface and touch screen allow rapid familiarization
  • Predefined and custom methods provide the results you need without the hassle of setup
  • PIN-protected user management functions provide security for compliance with regulatory agencies
These multiparameter meters feature an interface designed to improve work flow. The large touch screen makes navigation easy and the menu structure uses plain language, eliminating confusion during set up. One click will start one of 17 predefined or 60 custom methods, enhancing efficiency and throughput. Select the display that works best for you—uFocus™ shows only the most important data while the standard view shows more detail including setup parameters and measurements.
Security features help you comply with regulatory agency requirements. User management settings and PIN-protected functions prevent tampering with custom methods. Integrated Sensor Management (ISM®) pH electrodes and conductivity cells are automatically recognized by the meter and store their own calibration data ensuring every measurement is accurate even if you switch cells. Time and date stamps from the radio-controlled clock ensure measurements conform to GLP guidelines.
Included electrode arm smoothly lowers and lifts probes vertically with one hand, reducing spills and time between measurements. Modular construction allows you to expand your measurement capabilities as your lab needs evolve. Connect with PCs and peripheral devices using the USB, Ethernet, and RS-232 interfaces.
$3,290.00 - $4,630.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo SevenExcellence S470 Benchtop pH/Conductivity Meter Only EW-41000-13 Mfr # 30046252
COMPARE
Benchtop pH/Conductivity Meter Only
$3,290.00USD / Each
Mettler Toledo SevenExcellence S470 Benchtop pH/Conductivity Meter Standard Kit EW-41000-14 Mfr # 30046253
COMPARE
Benchtop pH/Conductivity Meter Standard Kit
$4,170.00USD / Each
Mettler Toledo SevenExcellence S470 Benchtop pH/Conductivity Meter USP/EP Kit EW-41000-15 Mfr # 30046254
COMPARE
Benchtop pH/Conductivity Meter USP/EP Kit
$4,630.00USD / Each
Mettler Toledo SevenExcellence S475 Benchtop pH/ISE/Conductivity Meter Only EW-41000-16 Mfr # 30046255
COMPARE
Benchtop pH/ISE/Conductivity Meter Only
$4,090.00USD / Each
Mettler Toledo SevenExcellence S470 Benchtop pH/Conductivity Meter Bio Kit EW-41000-17 Mfr # 64091338
COMPARE
Benchtop pH/Conductivity Meter Bio Kit
$4,440.00USD / Each
Showing 1- 5 of 5
Need Help with this product?
Call Us At1-847-549-7600
Live chat