METTLER TOLEDO SevenExcellence™ S700 Benchtop Conductivity Meters
So easy to operate—no need to study the manual to begin taking measurements
  • Intuitive plain-language interface and touch screen allow rapid familiarization
  • Predefined and custom methods provide the results you need without the hassle of setup
  • PIN-protected user management functions provide security for compliance with regulatory agencies
These conductivity meters feature an interface designed to improve work flow. The large touch screen makes navigation easy and the menu structure uses plain language, eliminating confusion during set up. One click will start one of 17 predefined or 60 custom methods, enhancing efficiency and throughput. Select the display that works best for you—uFocus™ shows only the most important data while the standard view shows more detail including setup parameters and measurements.
Security features help you comply with regulatory agency requirements. User management settings and PIN-protected functions prevent tampering with custom methods. Integrated Sensor Management (ISM®) conductivity cells are automatically recognized by the meter and store their own calibration data ensuring every measurement is accurate even if you switch cells. Time and date stamps from the radio-controlled clock ensure measurements conform to GLP guidelines.
Included electrode arm smoothly lowers and lifts the conductivity cell vertically with one hand, reducing spills and time between measurements. Modular construction allows you to expand your measurement capabilities as your lab needs evolve. Connect with PCs and peripheral devices using the USB, Ethernet, and RS-232 interfaces.
$3,355.00 - $3,920.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo S700 SevenExcellence Benchtop Conductivity Meter Only EW-41000-06 Mfr # 30046244
COMPARE
Benchtop Conductivity Meter Only
$3,355.00USD / Each
Mettler Toledo S700 SevenExcellence Benchtop Conductivity Meter Standard Kit EW-41000-07 Mfr # 30046245
COMPARE
Benchtop Conductivity Meter Standard Kit
$3,920.00USD / Each