This product is now discontinued, View Related Products
MicroDock II Automatic Test and Calibration Systems
Quick and easy compliance — automatically maintain accurate records
Manage the calibration and bump testing of your GasAlert portable gas detectors with
these calibration systems. Simplifying the record keeping process, the MicroDock II creates
accurate and user-friendly reports, prints receipts of calibration, sorts and graphs data,
and archives information. Results are stored on the MMC card for easy download to your
computer through the USB interface.
Also features automatic verification of audible and visual alarms. Calibrate multiple units
by adding the appropriate docking modules (up to 10). Connect multiple MicroDock II
systems via a LAN.
Call For Price
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
BW Technologies Microdock II Base Station w/GasAlertClip Extreme H2S Module EW-86419-51 Mfr # DOCK2-2-1E-00-G
COMPARE
Base Station w/GasAlertClip Extreme H2S Module
Call For Price
BW Technologies Microdock II Base Station w/GasAlertClip Extreme CO Module EW-86419-52 Mfr # DOCK2-2-1G-00-G
COMPARE
Base Station w/GasAlertClip Extreme CO Module
Call For Price
BW Technologies Microdock II Base Station w/GasAlertClip Extreme SO2 Module EW-86419-53 Mfr # DOCK2-2-1F-00-G
COMPARE
Base Station w/GasAlertClip Extreme SO2 Module
Call For Price
BW Technologies Microdock II Base Station w/GasAlertClip Extreme O2 Module EW-86419-54 Mfr # DOCK2-2-1H-00-G
COMPARE
Base Station w/GasAlertClip Extreme O2 Module
Call For Price
BW Technologies Microdock II Base Station w/GasAlert Extreme module EW-86419-55 Mfr # DOCK2-2-1D-00-G
COMPARE
Base Station w/GasAlert Extreme module
Call For Price
BW Technologies Microdock II Base Station w/GasAlertQuattro and Charger Module EW-86419-56 Mfr # DOCK2-2-1C1N-00-G
COMPARE
Base Station w/GasAlertQuattro and Charger Module
Call For Price
BW Technologies Microdock II Base Station w/GasAlertMicroClip module w/charger EW-86419-57 Mfr # DOCK2-2-1C1L-00-G
COMPARE
Base Station w/GasAlertMicroClip module w/charger
Call For Price
BW Technologies Microdock II Base Station w/GasAlertMax XT Module and Charger EW-86419-58 Mfr # DOCK2-2-1C1M-00-G
COMPARE
Base Station w/GasAlertMax XT Module and Charger
Call For Price
BW Technologies Microdock II Base Station w/GasAlertMicro 5 Module, Charger, 2-Port EW-86419-59 Mfr # DOCK2-2-1C1K-00-G
COMPARE
Base Station w/GasAlertMicro 5 Module, Charger, 2-Port
Call For Price
BW Technologies Microdock II Base Station w/GasAlertMicro 5 Module, Charger, 3-Port EW-86419-60 Mfr # DOCK2-3-1C1K-00-G
COMPARE
Base Station w/GasAlertMicro 5 Module, Charger, 3-Port
Call For Price
BW Technologies Microdock II GasAlertClip Extreme H2S Docking Module EW-86419-61 Mfr # DOCK2-0-1E-00-G
COMPARE
GasAlertClip Extreme H2S Docking Module
Call For Price
BW Technologies Microdock II GasAlertClip Extreme CO Docking Module EW-86419-62 Mfr # DOCK2-0-1G-00-G
COMPARE
GasAlertClip Extreme CO Docking Module
Call For Price
BW Technologies Microdock II GasAlertClip Extreme SO2 Docking Module EW-86419-63 Mfr # DOCK2-0-1F-00-G
COMPARE
GasAlertClip Extreme SO2 Docking Module
Call For Price
BW Technologies Microdock II GasAlertClip Extreme O2 Docking Module EW-86419-64 Mfr # DOCK2-0-1H-00-G
COMPARE
GasAlertClip Extreme O2 Docking Module
Call For Price
BW Technologies Microdock II GasAlertQuattro Docking Module, and Charger EW-86419-65 Mfr # DOCK2-0-1C1N-00-G
COMPARE
GasAlertQuattro Docking Module, and Charger
Call For Price
BW Technologies Microdock II GasAlertMicroClip Docking Module, and Charger EW-86419-66 Mfr # DOCK2-0-1C1L-00-G
COMPARE
GasAlertMicroClip Docking Module, and Charger
Call For Price
BW Technologies Demand Flow Regulator for Microdock II Base Station EW-86419-67 Mfr # REG-DF-1
COMPARE
Demand Flow Regulator for Microdock II Base Station
Call For Price
BW Technologies Flow Regulator with CGA connector for Microdock II EW-86419-68 Mfr # REG-DF-2
COMPARE
Flow Regulator with CGA connector for Microdock II
Call For Price
BW Technologies Flow Regulator for Disposable Cylinders; for Microdock II EW-86419-69 Mfr # REG-DF-3
COMPARE
Flow Regulator for Disposable Cylinders; for Microdock II
Call For Price
Showing 1- 19 of 19