Microflex® Diamond Grip Latex Exam Gloves
Proven performance that consistently exceeds the standards
  • Medical examination grade protects hands of healthcare professionals
  • Double chlorinated for easy donning and a comfortable feel
  • Chemical resistant
  • Powder-free to reduce skin irritation
  • Meets acceptable quality level (AQL) of 1.5
The gloves are designed to be extra thick at the fingers (7.9 mil) for added protection against rips, snags, and tears. Protects your hands in the most demanding environments. Textured fingers allow you to securely grip tools and materials — wet or dry. Natural latex ensures superior comfort and resilience.
The gloves are ideal for use in the automotive, lab research, life sciences, machinery, equipment and healthcare industries.
Standards and certifications include ASTM D3578, EN 420-2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, and US QSR/FDA 510(k) Medical Examination Grade. Ambidextrous.
$179.25 - $212.75USD / Case of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Palm Thickness (mil) Size
Availability Pricing
Microflex MF300-S Latex Gloves, Small; 1000/CS EW-86231-35 Mfr # MF300-S
COMPARE
9 1/2 6.3 Small
$183.75USD / Case of 1000
Microflex MF300-M Latex Gloves, Medium; 1000/CS EW-86231-36 Mfr # MF300-M
COMPARE
9 1/2 6.3 Medium
$179.25USD / Case of 1000
Microflex MF300-L Latex Gloves, Large; 1000/CS EW-86231-37 Mfr # MF300-L
COMPARE
9 1/2 6.3 Large
$183.75USD / Case of 1000
Microflex MF300-XL Latex Gloves, X-Large; 1000/CS EW-86231-38 Mfr # MF300-XL
COMPARE
9 1/2 6.3 X-Large
$212.75USD / Case of 1000