This product is now discontinued, View Related Products

Microflex FreeForm SE Nitrile Gloves 64121

Unique ''super soft'' formulation feels like latex
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.

Powder-free, 100% nitrile gloves offer superior comfort and elasticity in a synthetic glove. Unique formulation conforms to the contours of your hands as you move — minimizing unnecessary hand stress and enabling a full range-of-motion.

Lightly textured fingers ensure a secure grip in wet and dry conditions. Interior polymer coating assists in quick and easy donning.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
Microflex FFS700-S Nitrile Gloves, small, 1000/cs EW-86445-20 Mfr # FFS700-S
COMPARE
9 1/2 Small
Call For Price
Microflex FFS700-M Nitrile Gloves, medium, 1000/cs EW-86445-21 Mfr # FFS700-M
COMPARE
9 1/2 Medium
Call For Price
Microflex FFS700-L Nitrile Gloves, large, 1000/cs EW-86445-22 Mfr # FFS700-L
COMPARE
9 1/2 Large
Call For Price
Microflex FFS700-XL Nitrile Gloves, X-large, 1000/cs EW-86445-23 Mfr # FFS700-XL
COMPARE
9 1/2 X-Large
Call For Price
Microflex FreeForm SE Nitrile Gloves, extended cuff, small, 500/cs EW-86445-24 Mfr # FFE-775-S
COMPARE
11 1/2 Small
Call For Price
Microflex FreeForm SE Nitrile Gloves, extended cuff, medium, 500/cs EW-86445-25 Mfr # FFE-775-M
COMPARE
11 1/2 Medium
Call For Price
Microflex FreeForm SE Nitrile Gloves, extended cuff, large, 500/cs EW-86445-26 Mfr # FFE-775-L
COMPARE
11 1/2 Large
Call For Price
Microflex FreeForm SE Nitrile Gloves, extended cuff, extra large, 500/cs EW-86445-27 Mfr # FFE-775-XL
COMPARE
11 1/2 X-Large
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat