This product is now discontinued, View Related Products
Microflex XCEED Nitrile Gloves 64124
Value packed with more than double the normal amount of gloves!
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
New powder-free nitrile formulation provides strength and durability for all applications. The increased strength allows decreased thickness for added comfort and enhanced tactile sensitivity needed in laboratory applications. Same size box contains more than twice the normal amount of gloves — saving you money, time, and storage space. Fingertips are textured for a more secure grip; beaded cuff. Disposable; for single-use only. Ambidextrous.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Length (in) Size
Availability Pricing
Microflex XCEED Powder-Free Nitrile Glove, Extra small, 2500/cs EW-86445-35 Mfr # XC-310-XS
COMPARE
9 1/4 X-Small
Call For Price
Microflex XCEED Powder-Free Nitrile Glove, Small, 2500/cs EW-86445-36 Mfr # XC-310-S
COMPARE
9 1/4 Small
Call For Price
Microflex XCEED Powder-Free Nitrile Glove, Medium, 2500/cs EW-86445-37 Mfr # XC-310-M
COMPARE
9 1/4 Medium
Call For Price
Microflex XCEED Powder-Free Nitrile Glove, Large, 2500/cs EW-86445-38 Mfr # XC-310-L
COMPARE
9 1/4 Large
Call For Price
Microflex XCEED Powder-Free Nitrile Glove, Extra Large, 2300/cs EW-86445-39 Mfr # XC-310-XL
COMPARE
9 1/4 X-Large