This product is now discontinued, View Related Products
MicroPlate and Multi-MicroPlate Genie Shakers
High speed action for increased mixing efficiency in every well
  • Speed and orbit combine to offer true vortexing action in each well
  • Small vortexing orbit of 1.0 mm for thorough mixing regardless of sample viscosity
These vortexing shakers feature a built-in timer that operates from 1 to 60 minutes or continuous for hands-free operation. Mixing speed from 750 to 3200 rm for complete and uniform mixing in any microplate format. Accepts most microplate types within the recommendations of the Society for Biomolecular Screening (SBS), even in 384-well formats. Choose the 51402-24 or -26 to mix up to 4 microplates. Or add the optional accossory tray, 51402-27, to the -24 or -26 for mixing up to 8 total microplates.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Depth (in) Max Load (lbs) Width (in)
Availability Pricing
Scientific Industries Genie SI-0400 MicroPlate Shaker, 750 to 3200 rpm, 120 VAC EW-51402-20 Mfr # SI-0400A
COMPARE
5.75 1.1 7
Call For Price
Scientific Industries Genie SI-0401 MicroPlate Shaker, 750 to 3200 rpm, 240 VAC EW-51402-22 Mfr # SI-0401
COMPARE
5.75 1.1 7
Call For Price
Scientific Industries Genie SI-4000 Multi-MicroPlate , 750 to 3200 rpm, 120 VAC EW-51402-24 Mfr # SI-4000
COMPARE
13.2031 1.1 7
Call For Price
Scientific Industries Genie SI-4001 Multi-MicroPlate , 750 to 3200 rpm, 240 VAC EW-51402-26 Mfr # SI-4001
COMPARE
13.2031 1.1 7